Aluminium potassium sulphate dodecahydrate 98.0-102.0%, granular, AR® ACS, Macron Fine Chemicals™

Supplier: Avantor Performance Materials

Synonyms: Aluminium Potassium sulphate, 12-Hydrate, Aluminum potassium sulphate dodecahydrate, Aluminium potassium bis(sulphate) dodecahydrate, Potassium alum, Alum potassium

MACR3216-20EA 0 SGD
MACR3216-20
Aluminium potassium sulphate dodecahydrate 98.0-102.0%, granular, AR® ACS, Macron Fine Chemicals™
Aluminium potassium sulphate dodecahydrate
Formula: AlK(SO₄)₂·12H₂O
MW: 474.39 g/mol
Melting Pt: 92.5 °C
Density: 1.72 g/cm³ (20 °C)
MDL Number: MFCD00149143
CAS Number: 7784-24-9
EINECS: 233-141-3
Merck Index: 12,00373

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use 98.0 - 102.0 %
ACS - Assay (AlK(SO₄)₂·12H₂O) 98.0 - 102.0 %
ACS - Ammonium (NH₄) ≤0.005 %
ACS - Chloride (Cl) ≤5 ppm
ACS - Heavy Metals (as Pb) ≤0.0005 %
ACS - Insoluble Matter ≤0.005 %
ACS - Iron (Fe) ≤0.001 %
ACS - pH of 5% Solution at 25°C 2.0 - 4.0
ACS - Sodium (Na) ≤0.02 %
Solubility Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR