Ammonium carbonate ≥30% (NH₃ basis), AR® ACS, Macron Fine Chemicals™

Supplier: Avantor Performance Materials

Synonyms: Salt of hartshorn

MACR3352-20EA 0 SGD
MACR3352-20
Ammonium carbonate ≥30% (NH₃ basis), AR® ACS, Macron Fine Chemicals™
Ammonium carbonate
Formula: (NH₄)₂CO₃
MW: 96,09 g/mol
Melting Pt: 58 °C
Density: 1,5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010890
CAS Number: 10361-29-2
EINECS: 208-058-0
Merck Index: 13,00508

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use = 30 %
ACS - Assay (as NH₃) ≥30 %
ACS - Chloride (Cl) ≤5 ppm
ACS - Heavy Metals (as Pb) ≤5 ppm
ACS - Insoluble Matter ≤0.005 %
ACS - Iron (Fe) ≤5 ppm
ACS - Nonvolatile Matter ≤0.01 %
ACS - Sulfur Compounds (as SO₄) ≤0.002 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR