Copper(II) sulphate pentahydrate 98,0-102,0 Iodometry, crystals_fine, AR® ACS, Macron Fine Chemicals™

Supplier: Avantor Performance Materials
Danger

Synonyms: Cupric sulphate pentahydrate, Copper sulphate pentahydrate

MACR4844-04EA 0 SGD
MACR4844-04
Copper(II) sulphate pentahydrate 98,0-102,0 Iodometry, crystals_fine, AR® ACS, Macron Fine Chemicals™
Copper (II) sulphate pentahydrate

  • Not for pesticide use

Formula: CuSO₄·5H₂O
MW: 249.69 g/mol
Density: 2.284 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149681
CAS Number: 7758-99-8
EINECS: 231-847-6
UN: 3288
ADR: 6.1 III
REACH: 01-2119520566-40
Merck Index: 13,02682

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements 98.0 - 102.0%
For Laboratory,Research,or Manufacturing Use ≤0.001%
ACS - Assay (CuSO₄·5H₂O) (by iodometry) 98.0 - 102.0 %
ACS - Appearance (blue, crystalline granules) Passes Test
Solubility (5 in 50) Passes Test
ACS - Chloride (Cl) ≤0.001 %
ACS - Insoluble Matter ≤0.005 %
ACS - Iron (Fe) ≤0.003 %
ACS - Nitrogen Compounds (as N) ≤0.002 %
ACS - Calcium (Ca) ≤0.005 %
ACS - Nickel (Ni) ≤0.005 %
ACS - Potassium (K) ≤0.01 %
ACS - Sodium (Na) ≤0.02 %
Not for Pesticide Use

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR