Iron(II) sulphate, (dried basis) ≥97.0% (Fe basis), fine powder USP, FCC, Macron Fine Chemicals™

Supplier: Avantor Performance Materials
Warning

Synonyms: Ferrous sulphate

MACR5051-30EA 0 SGD
MACR5051-30
Iron(II) sulphate, (dried basis) ≥97.0% (Fe basis), fine powder USP, FCC, Macron Fine Chemicals™
Iron (II) sulphate
Formula: FeO₄S
MDL Number: MFCD00011014
CAS Number: 7720-78-7

Order Now

Specification Test Results

Meets F.C.C. Requirements Passes Test
Meets U.S.P Requirements Passes Test
Food GMP Manufactured Product ≤3 ppm
GMP Manufactured Product ≤2 mg/kg
CAUTION: For Manufacturing, processing or repackaging ≤1 mg/kg
Bulk Pharmaceutical Chemical 86.0 - 89.0%
Bulk Food Chemical Passes Test
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Arsenic (As) ≤3 ppm
USP - Lead (Pb) ≤2 mg/kg
USP - Mercury (Hg) ≤1 mg/kg
USP - Assay (as FeSO4) 86.0 - 89.0 %
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Insoluble Substances ≤0.05 %
FCC - Lead (Pb) ≤2 mg/kg
FCC - Mercury (Hg) ≤1 mg/kg
FCC - Assay (as FeSO4) 86.0 - 89.0 %
Mesh - Thru U.S. No. 40 Sieve ≥97.0 %
Mesh - Thru U.S. No. 100 Sieve ≥85.0 %
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Metallic Residues: No metal catalysts or metal reagents, as defined by
EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of
this material.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR