Hexane (mixture of isomers) ≥98.5% (by GC), AR® ACS, Macron Fine Chemicals™

Supplier: Avantor Performance Materials
Danger

MACR5189-26EA 4250 SGD
MACR5189-26 MACR5189-06 MACR5189-02 MACR5189-08 MACR5189-16 MACR5189-22
Hexane (mixture of isomers) ≥98.5% (by GC), AR® ACS, Macron Fine Chemicals™
Hexane (mixture of isomers)
Formula: H₃C(CH₂)₄CH₃
Density: 0,66 g/cm³ (20 °C)
Flash Pt: –23 °C
Storage Temperature: Ambient
MDL Number: MFCD02179311
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c38497992e162315d0f3563bfcd3dc478422095&vid=7880d919bf16a88cca44241139a2be8054e77804&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements ≥98.5%
For Laboratory,Research,or Manufacturing Use ≤10
ACS - Assay (sum of 5 isomers, total hexanes, plus methylcyclopentane)(by GC) ≥98.5 %
ACS - Assay (as n-hexane) (by GC), For Information Only ≤0.005%
Appearance (clear, colorless liquid) Passes Test
ACS - Color (APHA) ≤10
ACS - Residue after Evaporation ≤0.001 %
ACS - Sulfur Compounds (as S) ≤0.005 %
ACS - Thiophene Passes Test
ACS - Water-Soluble Titrable Acid (meq/g) ≤0.0003


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR