Cyclohexane, AR®, Laboratory Reagent, Macron Fine Chemicals™

Supplier: Avantor Performance Materials

Synonyms: Hexahydrobenzene, Hexamethylene

MACR5412-25EA 97.7 SGD
MACR5412-25
Cyclohexane, AR®, Laboratory Reagent, Macron Fine Chemicals™
Cyclohexane
Formula: C₆H₁₂
MDL Number: MFCD00003814
CAS Number: 110-82-7
UN: 1145
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e51ba998bef126804c3fd25b361be72cd9545&vid=d1e7f75d9f2a944eb15a7740c5254588cc9a4e67&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR