Methyl ethyl ketone, AR®, Laboratory Reagent, Macron Fine Chemicals™

Supplier: Avantor Performance Materials

Synonyms: EK 500 Additive, Ethyl methyl ketone, 2-Butanone, Butanone, MEK

MACR5447-25EA 0 SGD
MACR5447-25
Methyl ethyl ketone, AR®, Laboratory Reagent, Macron Fine Chemicals™
Methyl ethyl ketone
Formula: C₂H₅COCH₃
MDL Number: MFCD00011648
CAS Number: 78-93-3
UN: 1193
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b6656888f6a9a4c0c5ed3053bb2a3dcd8b3af66&vid=17cb98abf677b90f8bac752664d5c0cfd6765591&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR