Magnesium sulphate heptahydrate 99.5-100.5% (excluding ignited substances) USP, FCC, Macron Fine Chemicals™

Supplier: Avantor Performance Materials

Synonyms: sulphuric acid magnesium salt heptahydrate, Epsom salt, Bitter salt

Management of Change (MOC) category = R

MACR6046-24EA 0 SGD
MACR6046-24
Magnesium sulphate heptahydrate 99.5-100.5% (excluding ignited substances) USP, FCC, Macron Fine Chemicals™
Magnesium sulphate heptahydrate
Formula: MgSO₄·7H₂O
MW: 246.48 g/mol
Melting Pt: 1124 °C
Density: 1.67 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149785
CAS Number: 10034-99-8
EINECS: 231-298-2
Merck Index: 13,05715

Order Now

Specification Test Results

Food GMP Manufactured Product
GMP Manufactured Product
Meets F.C.C. Requirements
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
Bulk Food Chemical
USP - Assay (MgSO₄) (ignited basis) 99.0 - 100.5 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - pH (1 in 20) 5.0 - 9.2
USP - Loss on Ignition 40.0 - 52.0 %
USP - Chloride (Cl) ≤ 0.014 %
USP - Selenium (Se) ≤ 0.003 %
USP - Iron (Fe) ≤ 0.002 %
FCC - Assay (MgSO₄) (ignited basis) 99.5 - 100.5 %
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Lead (Pb) ≤ 4 mg/kg
FCC - Loss on Ignition 40.0 - 52.0 %
FCC - Selenium (Se) ≤ 0.003 %
Solubility (5 in 50) Passes Test
Appearance (small colorless crystals) Passes Test
Not intended for parenteral dosage use
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on
elemental impurities for this product is available on the
associated Product Regulatory Data Sheet and elemental impurity
profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR