Potassium permanganate 1.0 N aqueous solution, StandARd® Volumetric, Macron Fine Chemicals™

Supplier: Avantor Performance Materials

Synonyms: Permanganic acid potassium salt

MACR6139-01EA 0 SGD
MACR6139-01
Potassium permanganate 1.0 N aqueous solution, StandARd® Volumetric, Macron Fine Chemicals™
Potassium permanganate
Formula: KMnO₄
MW: 158.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS Number: 7722-64-7
EINECS: 231-760-3
UN: 3082
ADR: 9 III
Merck Index: 12,07824

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use 0.995 - 1.005
Normality 0.995 - 1.005

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR