n-Hexane 95%, ChromAR®, Macron Fine Chemicals™

Supplier: Avantor Performance Materials
Danger

Synonyms: Hexyl hydride, Hexanen, Hexane

MACR6752-25EA 94.6 SGD
MACR6752-25
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
n-Hexane
Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 69
Melting Pt: -94.3
Flash Pt: -22
MDL Number: MFCD00009520
CAS Number: 110-54-3
UN: 1208
ADR: 3 II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR