Ethyl acetate, AR®, Laboratory Reagent, Macron Fine Chemicals™

Supplier: Avantor Performance Materials

Synonyms: Acetic acid ethyl ester

MACR6780-25EA 67 SGD
MACR6780-25
Ethyl acetate, AR®, Laboratory Reagent, Macron Fine Chemicals™
Ethyl acetate
Formula: CH₃COOC₂H₅
MW: 88.11 g/mol
Boiling Pt: 77.1
Melting Pt: -83
Flash Pt: -4
MDL Number: MFCD00009171
CAS Number: 141-78-6
UN: 1173
ADR: 3 II
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR