Potassium hydrogen sulphate, powder_fused, AR® ACS, Macron Fine Chemicals™

Supplier: Avantor Performance Materials
Danger

Synonyms: Potassium bisulphate

MACR7112-06EA 0 SGD
MACR7112-06 MACR7112-20
Potassium hydrogen sulphate, powder_fused, AR® ACS, Macron Fine Chemicals™
Potassium hydrogen sulphate
Formula: KHSO₄
MW: 136.17 g/mol
Melting Pt: 214
Density: 2.32 g/cm³ (20 °C)
MDL Number: MFCD00011404
CAS Number: 7646-93-7
EINECS: 231-594-1
UN: 2509
ADR: 8 II
Merck Index: 13,07695

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use 37.5 - 38.6 %
ACS - Acidity (as H₂SO₄) 37.5 - 38.6 %
ACS - Water (H₂O) ≤2.5 %
ACS - Chloride (Cl) ≤0.002 %
ACS - Phosphate (PO₄) ≤0.001 %
ACS - Heavy Metals (as Pb) ≤0.001 %
ACS - Iron (Fe) ≤0.002 %
ACS - Sodium (Na) ≤0.01 %
ACS - Insoluble Matter ≤0.01 %
ACS - Ammonium Hydroxide Precipitate ≤0.01 %
ACS - Calcium (Ca) ≤0.002 %
ACS - Magnesium (Mg) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR