Sodium acetate trihydrate 99.0-101.0%, granular, AR® ACS, Macron Fine Chemicals™

Supplier: Avantor Performance Materials
Warning

Synonyms: Acetic acid sodium salt trihydrate

MACR7364-04EA 0 SGD
MACR7364-04 MACR7364-20 MACR7364-88 MACR7364-06
Sodium acetate trihydrate 99.0-101.0%, granular, AR® ACS, Macron Fine Chemicals™
Sodium acetate trihydrate

AR—The standard Macron Fine Chemicals™ grade of analytical reagents; suitable for laboratory and general use. If the reagent also meets the requirements of the American Chemical Society Committee on Analytical Reagents, it will be denoted as an AR (ACS) reagent. 

Formula: H₃CCOONa·3H₂O
MW: 136.08 g/mol
Boiling Pt: 120…123 °C (1013 hPa)
Melting Pt: 58
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00071557
CAS Number: 6131-90-4
EINECS: 204-823-8
Merck Index: 13,08642
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=313c1d45673fab192565649f8cbb7ddff555de39&vid=1ad37a966605626c95117735c83b16eaea381fbc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use 99.0 - 101.0 %
ACS - Assay (CH₃COONa·3H₂O) 99.0 - 101.0 %
ACS - Calcium (Ca) ≤0.005 %
ACS - Chloride (Cl) ≤0.001 %
ACS - Heavy Metals (as Pb) ≤5 ppm
ACS - Insoluble Matter ≤0.005 %
ACS - Iron (Fe) ≤5 ppm
ACS - Magnesium (Mg) ≤0.002 %
ACS - Phosphate (PO₄) ≤5 ppm
ACS - Potassium (K) ≤0.005 %
ACS - Substances Reducing Permanganate Passes Test
ACS - Sulfate (SO₄) ≤0.002 %
ACS - pH of 5% Solution at 25°C 7.5 - 9.2
Appearance (fine, colorless granules)
Product Information (not specifications):


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR