Copper(II) sulphate, blue 98,5-100,5, powder_crystalline, GenAR® USP Biotechnology, Macron Fine Chemicals™

Supplier: Avantor Performance Materials
Warning

Synonyms: Cupric sulphate

M.O.C. Category RL

MACR7790-88EA 0 SGD
MACR7790-88
Copper(II) sulphate, blue 98,5-100,5, powder_crystalline, GenAR® USP Biotechnology, Macron Fine Chemicals™
Copper (II) sulphate

Preserve in tight containers.

Formula: CuSO₄
MW: 159.61 g/mol
Melting Pt: 200
Density: 2.286 g/cm³ (15.6 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010981
CAS Number: 7758-98-7
EINECS: 231-847-6
UN: 3288
ADR: 6.1 III
Merck Index: 12,02722

Order Now

Specification Test Results

Meets U.S.P Requirements
Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
GMP Manufactured Product
Preserve in tight containers. Store at 25°C, excursions permitted 15-30°C. 98.5 - 100.5 %
CAUTION: For Manufacturing, processing or repackaging = 0.005 %
Bulk Pharmaceutical Chemical Passes Test
USP - Assay (as CuSO₄) 98.5 - 100.5 %
USP - Calcium (Ca) ≤0.005 %
Solubility (5 in 50) Passes Test
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Iron (Fe) ≤0.003 %
USP - Loss on Drying 33.0 - 36.5 %
USP - Nickel (Ni) ≤0.005 %
USP - Potassium (K) ≤0.01 %
USP - Sodium (Na) ≤0.02 %
Endotoxin Concentration (EU/g) ≤10
EP/BP - Assay 99.0 - 101.0 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Identification C Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Chloride (Cl) ≤100 ppm
EP/BP - Iron (Fe) ≤100 ppm
EP/BP - Lead (Pb) ≤50 ppm
EP/BP - Loss on Drying 35.0 - 36.5 %
Appearance (blue, crystalline powder)
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR