Sodium sulphate, anhydrous ≥99.0%, powder, AR® ACS, Macron Fine Chemicals™

Supplier: Avantor Performance Materials

Synonyms: Disodium sulphate, Bisodium sulphate, Sulphuric acid disodium salt

Management of Change (MOC) category = N

MACR8020-06EA 0 SGD
MACR8020-06
Sodium sulphate, anhydrous ≥99.0%, powder, AR® ACS, Macron Fine Chemicals™
Sodium sulphate

Powder.

Formula: Na₂SO₄
Boiling Pt: 1429 °C (1013 hPa)
Melting Pt: 888 °C
Density: 2,7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003504
CAS Number: 7757-82-6
EINECS: 231-820-9
Merck Index: 12,08829

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (Na₂SO₄) ≥99.0 %
ACS - Appearance (white, crystalline powder) Passes Test
Solubility and Foreign Matter Passes Test
ACS - Calcium (Ca) ≤0.01 %
ACS - Chloride (Cl) ≤0.001 %
ACS - Heavy Metals (as Pb) ≤5 ppm
ACS - Insoluble Matter ≤0.01 %
ACS - Iron (Fe) ≤0.001 %
ACS - Loss on Ignition ≤0.5 %
ACS - Magnesium (Mg) ≤0.005 %
ACS - Nitrogen Compounds (as N) ≤5 ppm
ACS - pH of 5% Solution at 25°C 5.2 - 9.2
ACS - Phosphate (PO₄) ≤0.001 %
ACS - Potassium (K) ≤0.002 %
Thru U.S. No. 50 Sieve ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR