L(+)-Ascorbic acid, fine powder USP, Macron Fine Chemicals™

Supplier: Avantor Performance Materials

Synonyms: Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum

MACR8829-03EA 0 SGD
MACR8829-03
L(+)-Ascorbic acid, fine powder USP, Macron Fine Chemicals™
L(+)-Ascorbic acid
Formula: C₆H₈O₆
MW: 176.13 g/mol
Boiling Pt: 553
MDL Number: MFCD00064328
CAS Number: 50-81-7
Merck Index: 13,00837
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c03499908e269560d680b0d7c4651f97a367aa0&vid=73a720d6900e713a98164022273a8afa6114ad7b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets U.S.P Requirements
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical 99.0 - 100.5 %
USP - Assay (C₆H₈O₆) 99.0 - 100.5 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Residue on Ignition ≤0.1 %
USP - Specific Rotation, [Å]²⁵^D (c=10 in water)(+) 20.5 - 21.5 Degree
Only Class 2 Solvents (Methanol) and Class 3 Solvents are likely to be
present. Residual Class 2 Solvents are below Option 1 and the residual concentration of all Class 3 Solvents is <0.5%.
Metallic Residues: No metal catalysts or metal reagents, as defined by
EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of
this material.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR