Sodium hexametaphosphate, Macron Fine Chemicals™

Supplier: Avantor Performance Materials

Synonyms: Sodium metaphosphate, SHMP

MACRE024-61EA 0 SGD
MACRE024-61
Sodium hexametaphosphate, Macron Fine Chemicals™
Sodium hexametaphosphate

Crystals.

Formula: Na₆P₆O₁₈
MW: 611,77 g/mol
Melting Pt: 610 °C
Density: 1,25 g/cm³ (20 °C)
MDL Number: MFCD00136045
CAS Number: 10124-56-8
EINECS: 233-343-1

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Phosphorus Pentoxide (P₂O₅) ≥65.0 %
pH of 1% Solution at 25°C 6.7 - 7.2
Appearance (clear, glass-like crystals)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR