1-Octanesulphonic acid sodium salt, monohydrate ≥98%, OR, Macron Fine Chemicals™

Supplier: Avantor Performance Materials
Warning

Synonyms: Sodium octane-1-sulphonate

MACRH165-56EA 0 SGD
MACRH165-56
1-Octanesulphonic acid sodium salt, monohydrate ≥98%, OR, Macron Fine Chemicals™
1-Octanesulphonic acid sodium salt

Powder. For use in PIC and HPLC applications.

Formula: C₈H₁₇NaO₃S
MW: 216.28 g/mol
Melting Pt: > 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007544
CAS Number: 5324-84-5
EINECS: 226-195-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1244cb11db07775dec2e193823e40080024b7036&vid=32d95cd9e321818e2f94c8261c4b4e6180ee0fc7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use = 98.0 %
Assay (CH₃(CH₂)₇SO₃Na·H₂O) ≥98.0 %
Solubility Passes Test
Optical Density at 210 nm (au) ≤0.05


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR