Macron Fine Chemicals™ SilicAR, Silica Gel 60 (230 - 400 Mesh)
Supplier: Avantor Performance Materials
MACRV120-01EA 0 SGD
MACRV120-01
Macron Fine Chemicals™ SilicAR, Silica Gel 60 (230 - 400 Mesh)
Chromatography Sorbents HPLC Sorbents
For column chromatography.
  • pH: 6,0 to 7,5
Packaging: Packed in mall HDPE wide mouth bottle.