Weighing paper, grade MN 226
Supplier: MACHEREY-NAGEL
186002
MANA186002EA 33.4 SGD
MANA186002
Weighing paper, grade MN 226
Weighing Papers
This is a transparent paper, smooth on both sides, which can be used as substitute for weighing boats. The smooth surface of the paper guarantees that the weighed goods can be transferred without loss.