Weighing boats, nitrogen-free parchment
Supplier: MACHEREY-NAGEL
486001
MANA486001EA 0 SGD
MANA486001
Weighing boats, nitrogen-free parchment
Boats Weighing Boats
Paper, nitrogen-free.
Weighing boats, MN 808 are used to weigh viscous or syrupy substances.