Qualitative filter papers, grade MN 617 wa
Supplier: MACHEREY-NAGEL
530018
MANA530018EA 0 SGD
MANA530018
Qualitative filter papers, grade MN 617 wa
Filters Filter Papers Qualitative Filter Paper
These papers are made hydrophobic (impermeable to water) by impregnation with a silicone. With the aid of these filters, water can be separated from water-immiscible organic solvents in an elegant manner, by means of a simple filtration.