Extraction thimbles
Supplier: MACHEREY-NAGEL
645001 645003 645008 645017 645024 645030 645041 645005 645022 645023 645031 645002 645004 645006 645007 645009 645010 645011 645013 645014 645015 645016 645018 645019 645020 645021 645025 645026 645027 645028 645029 645032 645033 645034 645035 645036 645037 645038 645039 645040 645042 645043 645220 645223 645225 645951
MANA645001EA 0 SGD
MANA645001 MANA645003 MANA645008 MANA645017 MANA645024 MANA645030 MANA645041 MANA645005 MANA645022 MANA645023 MANA645031 MANA645002 MANA645004 MANA645006 MANA645007 MANA645009 MANA645010 MANA645011 MANA645013 MANA645014 MANA645015 MANA645016 MANA645018 MANA645019 MANA645020 MANA645021 MANA645025 MANA645026 MANA645027 MANA645028 MANA645029 MANA645032 MANA645033 MANA645034 MANA645035 MANA645036 MANA645037 MANA645038 MANA645039 MANA645040 MANA645042 MANA645043 MANA645220 MANA645223 MANA645225 MANA645951
Extraction thimbles
Extractors Extraction Thimbles
Extraction thimbles, made of pure cellulose, standard quality. With or without lid available.
  • Fat determination in food and feed
  • Determination of binder content in asphalt
  • Analysis of soil contaminants
  • Determination of additives in rubber and elastomers