Guard column holder for preparative HPLC columns, VarioPrep
Supplier: MACHEREY-NAGEL
718251
MANA718251EA 0 SGD
MANA718251
Guard column holder for preparative HPLC columns, VarioPrep
Guard Column Holders
VarioPrep (VP) guard column holders are needed for the installation of VP guard columns at VP preparative HPLC columns.
  • Ideal protection for your preparative main column
  • VP column system with minimised void volume
VP guard column are packed with NUCLEODUR® or NUCLEOSIL® adsorbents for HPLC.