CHROMABOND® C18 ec SPE Columns
Supplier: MACHEREY-NAGEL
730141 730259 730404 730405 730011 730011ASP 730012 730012G 730013 730013ASP 730014 730014G 730015 730015ASP 730015G 731804 731805 731806 732012 732013 738011.100M 730015.250 730013.250 730013MPS 730014.250 730011MPS 730012MPS
MANA730141EA 0 SGD
MANA730141 MANA730259 MANA730404 MANA730405 MANA730011 MANA730011ASP MANA730012 MANA730012G MANA730013 MANA730013ASP MANA730014 MANA730014G MANA730015 MANA730015ASP MANA730015G MANA731804 MANA731805 MANA731806 MANA732012 MANA732013 MANA738011.100M MANA730015.250 MANA730013.250 MANA730013MPS MANA730014.250 MANA730011MPS MANA730012MPS
CHROMABOND® C18 ec SPE Columns
Chromatography Columns
CHROMABOND® C18 ec is an endcapped octadecyl modified silica phase for SPE.
  • Very nonpolar
  • Hydrophobic interactions with a wide variety of organic compounds
Suitable for nonpolar compounds as well as for aflatoxins, amphetamines, antibiotics, antiepileptics, barbiturates, caffeine, drugs, preservatives, fatty acids, nicotine, PAHs, pesticides, PCBs, heavy metals, and vitamins.
Order Now
SPECIFICATIONS
Environmentally Preferable
Carbon load 14%
Endcapped Yes Yes
pH range 2 - 8
Surface area 500 m²/g