SPE columns, CHROMABOND® C₆H₅
Supplier: MACHEREY-NAGEL
730084
MANA730084EA 0 SGD
MANA730084
SPE columns, CHROMABOND® C₆H₅
Chromatography Columns
CHROMABOND® C₆H₅ is a phenyl modified silica phase for SPE.
  • Polarity similar to C8 phase
  • In addition to hydrophobic interactions selective π-π interactions by the phenyl ring are possible
  • Especially suitable for aromatic substances
Recommended for aromatic compounds like phenols or nitroaromatics.