SPE columns, CHROMABOND® C2
Supplier: MACHEREY-NAGEL
730221
MANA730221EA 0 SGD
MANA730221
SPE columns, CHROMABOND® C2
Chromatography Columns
CHROMABOND® C2 is a dimethyl modified silica phase for SPE.
  • Very polar RP phase
  • Slight hydrophobic and strong polar interactions
  • Suitable for polar substances with hydrophobic areas from polar matrices
Recommended for biological molecules.