QuEChERS mixes, clean-up, CHROMABOND®
Supplier: MACHEREY-NAGEL
730972 730973 730974 730975 730358.2 730646.2 730657 730657.2 730658 730669 730850.2 730858.2 730996.2 730670.2 730842 730843.2 730845
MANA730972EA 0 SGD
MANA730972 MANA730973 MANA730974 MANA730975 MANA730358.2 MANA730646.2 MANA730657 MANA730657.2 MANA730658 MANA730669 MANA730850.2 MANA730858.2 MANA730996.2 MANA730670.2 MANA730842 MANA730843.2 MANA730845
QuEChERS mixes, clean-up, CHROMABOND®
Chromatography Columns
CHROMABOND® QuEChERS mixes with special diamino silica phase are suitable for determination of pesticides in food samples. These remove polar compounds (organic acids, pigments, sugars) from matrices like fruit or vegetables.
  • Quick, Easy, Cheap, Effective, Rugged, Safe
  • Multi-residue pesticide analysis of fruits or vegetables
  • Pre-weighed CHROMABOND® QuEChERS mixes
For subsequent clean-up, a diamino phase (PSA) removes sugars and organic acids. Magnesium sulphate removes water, C18 ec removes nonpolar interferences such as fats and the carbon phaseMore Product Information