SPE columns, cannabis analysis, CHROMABOND® Drug II
Supplier: MACHEREY-NAGEL
730680
MANA730680EA 0 SGD
MANA730680
SPE columns, cannabis analysis, CHROMABOND® Drug II
Chromatography Columns
CHROMABOND® Drug II is a special silica phase for the SPE of THC and its derivatives from biological fluids.
  • Base material: Silica
  • Special bifunctional modification: C8 / SB (strong anion exchanger)
Application: Extraction of THC and its derivatives from urine, blood, serum, and plasma as well as acidic analytes from biological fluids.