Vacuum manifold, CHROMABOND® MULTI 96
Supplier: MACHEREY-NAGEL
738630.M
MANA738630.MEA 0 SGD
MANA738630.M
Vacuum manifold, CHROMABOND® MULTI 96
Manifolds
CHROMABOND® MULTI 96 vacuum manifold is designed for applications of CHROMABOND® MULTI 96 SPE plates as well as for CHROMAFIL® MULTI 96 filter plates.
  • Suitable for 96-well monoblocks
  • Optimum flow rate due to vacuum adjustment by control valve
  • Reservoir tank is included and further accessories are available