Syringe filters, MN Steriliser CA
Supplier: MACHEREY-NAGEL
740401.50
MANA740401.50EA 0 SGD
MANA740401.50
Syringe filters, MN Steriliser CA
Filters Syringe Filters
MN steriliser CA syringe filter units are designed to sterilise and remove microorganisms and particulates from aqueous solutions such as biological fluids, culture media, additives, buffers, and protein solutions.
  • Low protein binding
  • Very robust membrane - high stability
  • Fast flow of liquid through the filter
  • Pre-sterilised with ethylene oxide
Housing material: Methyl methacrylate-butadiene-styrene polymer (MBS)