TLC plates, mixed layers, glass backing
Supplier: MACHEREY-NAGEL
810043
MANA810043EA 0 SGD
MANA810043
TLC plates, mixed layers, glass backing
TLC Plates
Glass plates with mixed layers for TLC.
Alox/CEL-AC-Mix-25: mixed layer of aluminium oxide G and acetylated cellulose recommended for separation of PAHs.
SILCEL-Mix-25: mixed layer of cellulose and silica, recommended for separation of preservatives and other antimicrobial compounds.
Order Now
SPECIFICATIONS
Environmentally Preferable