HPTLC plates, nano silica gel 60, modified layers, Nano-SIL DIOL
Supplier: MACHEREY-NAGEL
811120
MANA811120EA 0 SGD
MANA811120
HPTLC plates, nano silica gel 60, modified layers, Nano-SIL DIOL
TLC Plates
Diol-modified HPTLC silica layers available as glass plate with fluorescent indicator. Recommended for steroids, pesticides and plant constituents;
for critical separations an alternative to silica, since it is less sensitive to the water content of the environment; leads to more reproducible results compared to silica.
  • Layer can be wetted equally well with pure water as with organic solvents
Base material: Nano silica 60, specific surface (BET) ~ 500 m²/g; specific pore volume 0,75 mL/g; particle size 2 to 10 μm; pH stability 2 to 8.
Modification:More Product Information