TLC plates, silica gel 60, unmodified layers, DURASIL
Supplier: MACHEREY-NAGEL
812003 812004 812005 812006 812007 812008 812005.200
MANA812003EA 0 SGD
MANA812003 MANA812004 MANA812005 MANA812006 MANA812007 MANA812008 MANA812005.200
TLC plates, silica gel 60, unmodified layers, DURASIL
TLC Plates
These unmodified standard silica layers for TLC are available as glass plates with or without fluorescent indicator.
  • Hard, water-resistant and wettable layers due to a special binder system
  • More polar than SIL G
Silica 60: Specific surface (BET) ~ 500 m²/g; specific pore volume 0,75 ml/g; particle size 5 to 17 μm.
Order Now
SPECIFICATIONS
Environmentally Preferable