HPTLC Plates, Silica Gel 60, Modified Layers, RP-18 W/UV₂₅₄
Supplier: MACHEREY-NAGEL
811073 811075 811071 818144 818145 818146 818147 818152 811072 811074
MANA811073EA 0 SGD
MANA811073 MANA811075 MANA811071 MANA818144 MANA818145 MANA818146 MANA818147 MANA818152 MANA811072 MANA811074
HPTLC Plates, Silica Gel 60, Modified Layers, RP-18 W/UV₂₅₄
TLC Plates
Octadecyl-modified HPTLC silica layers available as glass plates or ALUGRAM® aluminium sheets. Recommended for aminophenol, barbiturates, preservatives, nucleobases, polycyclic aromatic, hydrocarbons, steroids, tetracyclines, and plasticisers (phthalates).
Base material: Nano-silica 60: Specific surface (BET) ~ 500 m²/g; specific pore volume 0,75 ml/g; particle size 2 to 10 μm; pH stability 2 to 10
Silica 60 for preparative plates (1000 µm): Specific surface (BET) ~ 500 m²/g; specific pore volume 0,75 ml/g; particle size 5 to 17 μm; pH stability 2 to 10
Modification: Partial octadecyl,More Product Information