HPTLC plates, nano silica gel 60, unmodified layers, Nano-SIL
Supplier: MACHEREY-NAGEL
811013 818141 818143 811011 811012 811021 811022 811023 818343 818240 818241 818342
MANA811013EA 0 SGD
MANA811013 MANA818141 MANA818143 MANA811011 MANA811012 MANA811021 MANA811022 MANA811023 MANA818343 MANA818240 MANA818241 MANA818342
HPTLC plates, nano silica gel 60, unmodified layers, Nano-SIL
TLC Plates
Unmodified HPTLC silica layers for HPTLC.
  • Nano silica gel (particle size 2 to 10 μm)
  • Narrow fractionation of the silica particles
  • Binders are stable in almost all organic solvents
Nano-SIL HPTLC plates and sheets allow sharper separations, shorter developing times and an increased detection sensitivity compared to standard silica gel plates.
Nano silica 60: Specific surface (BET) ~ 500 m²/g; specific pore volume 0,75 ml/g; particle size 2 to 10 μm.
Order Now
SPECIFICATIONS
Environmentally Preferable