TLC plates, silica gel 60, unmodified layers, ADAMANT
Supplier: MACHEREY-NAGEL
821005 821010 821015 821020 821025 821030 821040 821050 821060 821010.200 821040.200
MANA821005EA 0 SGD
MANA821005 MANA821010 MANA821015 MANA821020 MANA821025 MANA821030 MANA821040 MANA821050 MANA821060 MANA821010.200 MANA821040.200
TLC plates, silica gel 60, unmodified layers, ADAMANT
TLC Plates
These unmodified standard silica layers for TLC are available as glass plates with or without fluorescent indicator. High suitability for trace analyses resulting from a UV indicator with increased brilliance and a low-noise background of the layer.
  • Efficient hardness and abrasion resistance due to an optimised binder system
  • Increased separation efficiency due to an optimised particle size distribution
Silica 60: Specific surface (BET) ~ 500 m²/g; specific pore volume 0,75 ml/g; particle size 5 to 17 μm.