NANOCOLOR® Photometric Standard Test Kit for Cobalt
Supplier: MACHEREY-NAGEL
91851
MANA91851EA 0 SGD
MANA91851
NANOCOLOR® Photometric Standard Test Kit for Cobalt
Environmental Test Kits
Photometric test kit for the determination of cobalt in water samples.
  • Ready-to-use reagents for up to 500 determinations
  • Excellent sensitivity and accuracy due to measurements in 50 mm cuvettes
  • Low costs per determination
  • Easy dilutions for analysis of higher concentrations
The test kit is evaluated in 10 or 50 mm rectangular cuvettes. It is preprogrammend on NANOCOLOR® photometers. Programming data for other spectrophotometers are available.
Sea water analysis: Yes
Hazardous material: Yes
Remark: Measuring range on NANOCOLOR VIS II
Delivery information: Rugged box with reagents for 250 tests.