NANOCOLOR® Photometric Standard Test Kit for Copper

Supplier: MACHEREY-NAGEL

91853
MANA91853EA 0 SGD
MANA91853
NANOCOLOR® Photometric Standard Test Kit for Copper
Environmental Test Kits

Photometric test kit for the determination of copper in water samples.



  • Ready-to-use reagents for up to 500 determinations

  • Excellent sensitivity and accuracy due to measurements in 50 mm cuvettes
  • Low costs per determination
  • Easy dilutions for analysis of higher concentrations


The test kit is evaluated in 10 or 50 mm rectangular cuvettes. It is preprogrammend on NANOCOLOR® photometers and also compatible with METTLER TOLEDO Easy VIS and Easy UV. Programming data for other spectrophotometers are available.



Sea water analysis: Yes

Remark: Measuring range on NANOCOLOR VIS II



Delivery information: Rugged box with reagents for 250 tests.

Order Now

SPECIFICATIONS

Model MANA91853
Environmentally Preferable
Platform NANOCOLOR® standard tests
Storage temperature 15…25 °C
Shelf life (months) 24
Weight 574,9 g (including packaging)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR