NANOCOLOR® Photometric Standard Test Kit for Nitrite

Supplier: MACHEREY-NAGEL

91867
MANA91867EA 0 SGD
MANA91867
NANOCOLOR® Photometric Standard Test Kit for Nitrite
Environmental Test Kits

Photometric test kit for the determination of nitrite in water samples.



  • Ready-to-use reagents for up to 500 determinations

  • Excellent sensitivity and accuracy due to measurements in 50 mm cuvettes
  • Low costs per determination
  • Easy dilutions for analysis of higher concentrations


The test kit is evaluated in 10 or 50 mm rectangular cuvettes. It is preprogrammend on NANOCOLOR® photometers and also compatible with METTLER TOLEDO Easy VIS and Easy UV. Programming data for other spectrophotometers are available.



Sea water analysis: Yes
Hazardous material: Yes

Remark: Measuring range on NANOCOLOR VIS II


Order Now

SPECIFICATIONS

Model MANA91867
Environmentally Preferable
Platform NANOCOLOR® standard tests
Storage temperature 15…25 °C
Shelf life (months) 12
Weight 550,23 g including packaging

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR