NANOCOLOR® Photometric Tube Test Kit for Potassium

Supplier: MACHEREY-NAGEL

985045
MANA985045EA 0 SGD
MANA985045
NANOCOLOR® Photometric Tube Test Kit for Potassium
Environmental Test Kits

Photometric test kit for the determination of potassium in water samples.


  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Bar code cuvette identification for easy test call up in NANOCOLOR® spectrophotometers
  • Precisely pre-dosed reagents with constant high quality from batch to batch
  • Coloured pictogram instructions and easy handling of the tubes
  • 16 mm (OD) tubes for easy pipetting


The test kit is evaluated in 16 mm (OD) tubes. It is preprogrammend on NANOCOLOR® photometers and also compatible with METTLER TOLEDO Easy VIS and Easy UV. Programming data for other spectrophotometers are available.


Sea water analysis: Yes
Hazardous material: Yes
Remark: Measuring range on NANOCOLOR VIS II


Delivery information: Rugged box with 20 test tubes.

Order Now

SPECIFICATIONS

Model MANA985045
Environmentally Preferable
Platform NANOCOLOR® tube tests
Storage temperature 15…25 °C
Shelf life (months) 24
Weight 386.4 g (including packaging)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR