NANOCOLOR® Photometric Tube Test Kit for Silver
Supplier: MACHEREY-NAGEL
985049
MANA985049EA 0 SGD
MANA985049
NANOCOLOR® Photometric Tube Test Kit for Silver
Environmental Test Kits
A pre-dosed NANOCOLOR® tube test for photometric determination of silver in water.
  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Barcode cuvette identification
  • Precisely pre-dosed reagents in 16 mm (ext.Ø) tubes
The test is particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well.
Sea water analysis: No
Hazardous material: Yes
Remark: Measuring range on NANOCOLOR VIS II
Delivery information: Rugged box with 20 test tubes.