NANOCOLOR® Photometric Tube Test Kit for Methanol

Supplier: MACHEREY-NAGEL

985859
MANA985859EA 0 SGD
MANA985859
NANOCOLOR® Photometric Tube Test Kit for Methanol
Environmental Test Kits

A pre-dosed NANOCOLOR® tube test for photometric determination of methanol in water.


  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents

  • Barcode cuvette identification
  • Precisely pre-dosed reagents in 16 mm (ext.Ø) tubes


The test is particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well.


Sea water analysis: No

Hazardous material: No

Remark: Measuring range on NANOCOLOR VIS II



Delivery information: Rugged box with 23 test tubes.

Order Now

SPECIFICATIONS

Model MANA985859
Environmentally Preferable
Platform NANOCOLOR® tube tests
Storage temperature <0 °C
Shelf life (months) 24
Weight 573 g (including packaging)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR