Counting chamber, McMaster
Supplier: MARIENFELD
0611240
MARI0611240EA 0 SGD
MARI0611240
Counting chamber, McMaster
Slides Counting Chambers
This counting chamber is designed for applications with solid media.
  • Cover glass with two counting grids (10×10 mm, divided into 10 parts) affixed on three supports
  • Distance between bottom plate and cover glass (1,5 mm)
Suitable for counting worm eggs.