Counting chambers, Bürker
Supplier: MARIENFELD
0640211 0640210 0640231 0640331 0640310 0640311 0640330 0640230
MARI0640211EA 0 SGD
MARI0640211 MARI0640210 MARI0640231 MARI0640331 MARI0640310 MARI0640311 MARI0640330 MARI0640230
Counting chambers, Bürker
Slides Counting Chambers
Glass counting chambers equipped with two sets of grids, engraved into two ground, and polished surfaces on the central ridge. This central ridge is located between two elevated ridges, that are also ground and polished.
Designed for investigations of cell suspensions.
Conform to DIN 12847 and the German Calibration Ordinance
Certifications: DIN 12847 and the German Calibration Ordinance.
Delivery information: Supplied in a transparent box.
Order Now
SPECIFICATIONS
Environmentally Preferable