Counting chambers with V-slash, Imp. Neubauer
Supplier: MARIENFELD
0650010 0650030
MARI0650010EA 135 SGD
MARI0650010 MARI0650030
Counting chambers with V-slash, Imp. Neubauer
Slides Counting Chambers
These counting chambers with V-slash facilitates the feeding of the capillary gap between chamber bottom and haemacytometer cover glass with reduced risk of overflow.
Certifications: IVD 98/79/EC
Order Now
SPECIFICATIONS
Environmentally Preferable