Microscope slides
Supplier: MARIENFELD
1000000 1000200 1000412 1000612
MARI1000000EA 6.9 SGD
MARI1000000 MARI1000200 MARI1000412 MARI1000612
Microscope slides
Slides Microscope Slides
Slides designed for in vitro diagnostic (IVD) applications.
  • Pre-washed and ready to use
  • Either ground or cut versions
  • Soda-lime glass, Hydrolytic Class 3
Extra-white slides are used for low auto-fluorescence applications.