Cavity slides
Supplier: MARIENFELD
1320000 1320202 1320002 1320102
MARI1320000EA 0 SGD
MARI1320000 MARI1320202 MARI1320002 MARI1320102
Cavity slides
Slides Cavity Slides
Soda-lime glass of hydrolytic class 3 cavity slides.
Carefully ground and polished for in vitro diagnostic (IVD) applications.
Certifications: IVD 98/79/EC
Order Now
SPECIFICATIONS
Environmentally Preferable