Nessler cylinders, soda lime glass
Supplier: MARIENFELD
2800011 2800012 2800014
MARI2800011EA 0 SGD
MARI2800011 MARI2800012 MARI2800014
Nessler cylinders, soda lime glass
Tubes Nessler Tubes
Nessler cylinders are made of soda lime glass with white circular mark and spout. They are used for comparison of colour and turbidity between solutions.