Capillaries for melting point determination
Supplier: MARIENFELD
2930207 2930210 2930205 2940205 2930208 2940208 2930201 2930203 2930204 2940206 2940207 2940209 2940200 2940210 2940202
MARI2930207EA 296 SGD
MARI2930207 MARI2930210 MARI2930205 MARI2940205 MARI2930208 MARI2940208 MARI2930201 MARI2930203 MARI2930204 MARI2940206 MARI2940207 MARI2940209 MARI2940200 MARI2940210 MARI2940202
Capillaries for melting point determination
Tubes Melting Point Tubes and Capillaries
These single use, glass capillary tubes can be used for storage of small amounts of liquids or for the determination of melting point.
  • With rectangular cut edges
  • Tolerance length: ± 1 mm
  • Tolerance diameter: ± 0,05 mm
Type A capillaries have two open ends
Type B capillaries have one closed end
Order Now
SPECIFICATIONS
Environmentally Preferable